Tisk…

1-(Heptafluorobutyryl)imidazole ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®

Dodavatel: Merck

74382-10X1ML 74382-10ML 74382-1ML
74382-10X1MLEA 19760 CZK
74382-10X1ML 74382-10ML 74382-1ML
1-(Heptafluorobutyryl)imidazole ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®
1-(Heptafluorobutyryl)imidazole
Vzorec: C₇H₃F₇N₂O
MDL Number: MFCD00014503
CAS číslo: 32477-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4ebc844bc9f067b7a2d38d7c3f7a0ccf4e24c27&vid=ecc81daad25f151efaabf82d5d16963bf912c46b&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR