Tisk…

Chloroform, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

7386.2500EA 5790 CZK
7386.2500 7386.1000 7386.9025
Chloroform, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Chloroform
Vzorec: CHCl₃
Bod varu: 61,7 °C (1013 hPa)
Bod tání: –63 °C
Hustota: 1,4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS číslo: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Acetone and Aldehyde (as (CH3)2CO) max. 0.005 %
Acid and Chloride passes test
Density (g/ml) at 25°C 1.467 - 1.474
Free Chlorine passes test
Preservative (C2H5OH) about 0.75%
Substances Darkened by H2SO4 passes test
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1
Additional Specification(s):
Assay (by GC) min. 99 %
Color (APHA) max. 10
Residue after Evaporation max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR