Tisk…
Deepwell Plates 96-Well, 2000 µl
Dodavatel: EPPENDORF
0030501.306 0030501.314 0030501.330 0030501.349 0030502.302 0030502.310 0030502.337 0030502.345 0030505.301 0030506.308 0030113.527
737-2510EA 5850 CZK
737-2510 737-2511 737-2513 737-2514 737-2525 737-2526 737-2528 737-2529 737-0202 737-0205 737-0250
Deepwell Plates 96-Well, 2000 µl
Mikrodestičky Destičky s hlubokými jamkami
Eppendorf Deepwell plates 96-well, 2000 µl offer efficient sample handling with features like OptiTrack® matrix for rapid identification and g-Safe® technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample
More Product Information