Tisk…
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Dodavatel: EPPENDORF
0030128.648 0030128.680 0030128.672 0030128.664 0030128.656 0030128.850 0030128.842 0030128.796 0030128.788 0030128.770 0030133.331 0030129.318 0030133.307 0030129.300 0030133.358 0030133.382 0030129.601 0030129.849 0030129.857 0030113.560 0030113.616 0030113.624 0030113.632
732-0107EA 4310 CZK
732-0107 391-2054 732-0110 732-0109 732-0108 EPPE0030128.850 EPPE0030128.842 737-0164 EPPE0030128.788 EPPE0030128.770 731-0411 737-0180 731-0408 731-0414 731-0413 713-0021 EPPE0030129.601 EPPE0030129.849 EPPE0030129.857 737-0256 EPPE0030113.616 EPPE0030113.624 EPPE0030113.632
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Mikrodestičky Destičky pro PCR
Eppendorf's twin.tec® 96-well, 150 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. The SafeCode variants enhance sample identification, while the BioBased options, manufactured using renewable resources, provide an eco-friendly alternative without compromising performance.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid
More Product Information