Tisk…
Eppendorf LoBind® twin.tec® PCR Plates
Dodavatel: EPPENDORF
0030129.504 0030129.512 0030129.547 0030129.555 0030129.563 0030129.598 0030129.660 0030129.679
737-0185EA 5190 CZK
737-0185 737-0186 EPPE0030129.547 EPPE0030129.555 EPPE0030129.563 731-0705 731-0706 EPPE0030129.679
Eppendorf LoBind® twin.tec® PCR Plates
Mikrodestičky Destičky pro PCR
Eppendorf twin.tec® PCR plates LoBind, available in 96 and 384-well formats, maximize nucleic acid recovery and are certified as 'PCR clean' for consistently reliable results.
  • LoBind material guarantees maximum sample recovery for improved assay results
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer
More Product Information