Tisk…
Protein coated cell culture plates, CELLCOAT®, Poly-lysine
Dodavatel: Greiner Bio-One
781946_20 657930_50 662930_50 655944_20 781936_20 655946_20 655930_20 655936_20 655940_20 781940_20 781944_20 655948_120 662940_50
737-0080EA 22260 CZK
737-0080 392-0038 392-0045 392-0027 737-0079 392-0028 738-0012 738-0013 738-0014 392-0134 392-0135 392-0282 392-0277
Protein coated cell culture plates, CELLCOAT®, Poly-lysine
Mikrodestičky Destičky pro bunečné kultury
PS, coated with poly-lysine. Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic molecules that are used to improve the adhesion of different cell types to polystyrene surfaces. The cultivation efficiency of individual cell lines can be improved, especially when serum-free or serum-reduced medium is used, or when experiments such as transfections are performed. As the PDL is synthetic it is free of the impurities carried by other proteins.
  • Improved cell adhesion, proliferation and growth of transfected cell lines (eg. HEK-293, PC12, L929, certain 3T3
More Product Information