Tisk…
Microplate washer, Wellwash™ Versa
Dodavatel: THERMO LABSYSTEMS LIFE SCIENCE
5165010 5165050
736-0420EA 241950 CZK
736-0420 736-3009
Microplate washer, Wellwash™ Versa
Promývačky mikrodestiček
Wash 96- or 384-well plates securely with the Thermo Scientific™ Wellwash™ Versa Microplate Washer, an easy-to-use microplate-strip washer for routine ELISA applications. Designed for high performance and versatility, enjoy the ease of use and convenience provided through a graphical interface, local language version and USB port.
  • Easy and convenient to use
  • Large color display and keypad for easy, convenient use
  • Clear display shows everything on one screen
  • User interface is logic and intuitive, allowing quick protocol development
  • Easy-to-use software