Tisk…
U96 DeepWell™ plates 1,3 and 2,0 ml, Nunc®
Dodavatel: Thermo Fisher Scientific
260251 260252 278743 278752
736-0600EA 20410 CZK
736-0600 736-0601 736-0606 736-0607
U96 DeepWell™ plates 1,3 and 2,0 ml, Nunc®
Mikrodestičky Destičky s hlubokými jamkami
PP. Low design with shared wall technology for increased well volume. Ideal for sample collection, storage and combinatorial chemistry library applications.
  • Round well shape reduces liquid retention
  • Resistant to most chemicals, solvents and alcohols used in combinatorial chemistry
  • Well design for optimal storage capacity and increased mixing
  • Alphanumeric grid for quick sample identification
  • Sterile format certified free from RNase and DNase
Colour: Clear.
Colour: Clear