Tisk…
Microplates, 96- and 384-well, UV Star®
Dodavatel: Greiner Bio-One
655801_40 781801_40 675801_40 655809_40
736-0231EA 20010 CZK
736-0231 736-0153 738-0513 GREI655809_40
Microplates, 96- and 384-well, UV Star®
Mikrodestičky Mikrodestičky pro HTS
These microplates combine a unique process technology with a proprietary resin for superior UV transparency.
  • Improved chemical resistance
  • Free of detectable DNase, RNase, and human DNA
  • Non pyrogenic
  • Flat 135 µm film bottoms ensure exceptionally low backgrounds for more accurate measurements
  • 100% transmittance at preferred wavelengths
  • Innovative moulding technique eliminates possibility of film detachment
  • Suitable for storage down to –80 °C; DMSO resistant
Standard PS microplates are only partially suitable for UV transmission measurements, as PSMore Product Information