Tisk…
Immunoassay microplates
Dodavatel: Thermo Fisher Scientific
95029390 3355 3455 3555 3655 3855 6301 6302 6309 6310 6404 6405 9502227 9502887 95029100 95029140 95029180 95029330
735-0551EA 8560 CZK
735-0551 735-0461 735-0462 735-0463 735-0464 735-0465 735-0491 735-0492 735-0493 735-0494 735-0499 735-0500 735-0531 735-0535 735-0537 735-0539 735-0540 735-0546
Immunoassay microplates
Mikrodestičky Mikrodestičky a stripy pro imunologii
Designed specifically for immunoassay applications, this range of products have binding surfaces that include Immulon®, universal binding (UB) and enhanced binding (EB). These make good experimental design easier, allowing development of assays with stringent pH, buffer, pKa and resin requirements.
  • Available in 96-well or strip plate format for matching to the requirements of automated instruments
  • Microplates with breakable strips offer maximum efficiency for processing both large and small sample numbers, as the exact number of wells required can be used
More Product Information