Tisk…
MicroWell™ Plates, 96 well, Nunc-Immuno™
Dodavatel: Thermo Fisher Scientific
430341 439454 446140 446612 449824 456529 456537 460984 467320 467340 475094 475434 442404
735-0079EA 4830 CZK
735-0079 735-0199 735-0096 735-0097 735-0030 735-0033 735-0034 735-0038 735-0044 735-0045 735-0131 735-0068 735-0083
MicroWell™ Plates, 96 well, Nunc-Immuno™
Mikrodestičky Mikrodestičky pro HTS
PS, clear, without lid. Plates designed specifically for quantitative and qualitative solid phase immunoassays (for example, ELISA) and binding assays. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ plates are certified for binding homogeneity.
  • Choice of high flange or pinchbar design and flat (F), round (U) or C-shaped well bottoms
  • Plates have alphanumeric markings and fit all standard equipment
  • Barcoded version also available
MaxiSorp™ surface has a high affinity to molecules with mixedMore Product Information