Tisk…
Organic reference standard, Bisphenol A (2,3-dihydroxypropyl) glycidyl ether, analytical standard, Supelco®
Dodavatel: Merck
73417-100MG 73417-25MG
73417-100MGEA 16600 CZK
73417-100MG 73417-25MG
Organic reference standard, Bisphenol A (2,3-dihydroxypropyl) glycidyl ether, analytical standard, Supelco®
Standardy Standardy organické
Bisphenol A (2,3-dihydroxypropyl) glycidyl ether can be obtained as a product of the reaction of bisphenol A with epichlorohydrin.It is generally used as a substrate to prepare epoxy resins, which is used in coatings and adhesives.
Balení: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e19e5f81b838a76a197530b98f335e36fc0a0476&vid=e19e5f81b838a76a197530b98f335e36fc0a0476&prid=web&clid=SAPDAM