Tisk…
Spinner flasks, disposable, Corning®
Dodavatel: Corning
3152 3153 3561 3563 3580 3581 3578
734-4049EA 36700 CZK
734-4049 734-4050 734-4097 734-4099 734-2289 734-2290 392-0309
Spinner flasks, disposable, Corning®
Baňky Baňky míchané
The Corning® disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
  • Paddle size and height is optimised for each vessel size
  • Integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers
  • Heat build-up in the vessel is reduced by means of a specially designed flange that raised the vessel off the stir-plate surface for the 125 and 500 ml flasks only
Molded from virgin polystyrene and gamma-irradiated, each spinnerMore Product Information