Tisk…
Cell culture dishes, Nunc™ EasYDish™
Dodavatel: Thermo Fisher Scientific
150460 150462 150464 150466 150468
734-3254EA 6710 CZK
734-3254 734-3255 734-3256 734-3257 734-3258
Cell culture dishes, Nunc™ EasYDish™
Misky kultivační Misky pro tkáňové kultury
Thermo Scientific™ Nunc™ EasYDish™ cell culture dishes are designed to improve the handling, stacking, and transporting of cell cultures in the laboratory. Made of PS, USP Class VI they are free from RNase, DNase and human DNA, as well as non cytotoxic and non pyrogenic.
  • Beveled grip ring is easy to hold and stays secure in gloved hands
  • Proprietary notch design allows optimal ventilation for cell growth whilst maintaining sterility, minimises friction between the lid and rim, restricts movement of the lid to help secure lid-to-dish fitting, and increases
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí IRECS