Tisk…
Dialysis systems Spectra/Por® Float-A-Lyzer® G2
Dodavatel: Spectrum Laboratories
G235029 G235053 G235065 G235031 G235055 G235067 G235033 G235057 G235069 G235035 G235059 G235071 G235061 G235025 G235034 G235058 G235049 G235070 G235027 G235051 G235063 G235036 G235037 G235060 G235062 G235072 G235073
734-3573EA 6710 CZK
734-3573 734-3577 734-3581 734-3574 734-3578 734-3582 734-3575 734-3579 734-3583 734-3576 734-3580 734-3584 734-1498 734-1494 734-1495 734-1497 734-1496 734-1499 734-2363 734-2366 734-2606 734-2364 734-2365 734-2367 734-2368 734-2369 734-2370
Dialysis systems Spectra/Por® Float-A-Lyzer® G2
Hadičky Hadičky dializační
Ready-to-use laboratory dialysis systems featuring proprietary Ultra-pure Biotech Cellulose Ester (CE) Membrane. Biotech Grade CE is a low protein-binding synthetic membrane available in 9 precise MWCO's with no heavy metal and sulfide contaminants. The cylindrical tubing geometry prevents sample dilution (associated with cassette-type devices) and provides open access for total volume retrieval by pipette. Only the Float-A-Lyzer® G2 assures a 95 to 98% sample recovery while maintaining 99% sample purity and <1% sample dilution. The leakproof screw-on cap with sealing o-ring provides easyMore Product Information