Tisk…
Cell culture flasks, Corning®
Dodavatel: Corning
430372 430639 430720 430824 430825 431079 431080 431081 431082 431085 431306 430168 430823 3292 3290 3289 3291 3293 431328 3814 431463 431465 431466 430641U 430720U 430725U 431464U
734-1706EA 18990 CZK
734-1706 734-1712 734-1715 734-1718 734-1719 734-1722 734-1723 734-1724 734-1725 734-1726 734-1733 734-1700 734-1717 734-0093 734-0091 734-0090 734-0092 734-0094 734-1214 734-4139 734-2272 734-2274 734-2275 734-2705 734-2706 734-2707 734-2708
Cell culture flasks, Corning®
Baňky Baňky pro buněčné kultury
Manufactured from optically clear virgin polystyrene.
  • Printed with lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilised by gamma irradiation and certified non pyrogenic
  • Canted models have a skirt to prevent tipping
Non-treated: Flasks are designed for use in applications where cell attachment is not desired.
TC treated: For optimal cell attachment and growth.
Ultra low attachment surface: Features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation,More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí RECs, ISO 14001 - Oneonta