Tisk…

VWR®, Taq DNA polymerase, 200000 U

Dodavatel: VWR Chemicals

733-2479EA 0 CZK
733-2479
VWR®, Taq DNA polymerase, 200000 U
Činidla pro nukleové kyseliny End-point PCR Enzymes and Kits

Taq DNA polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from thermus aquaticus and has a molecular weight of approximately 94 kDa.


  • Store at –20 °C


Enzyme is supplied in 20 mM tris-HCl pH 8,5, 100 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5% tween 20, 0,5% NP40, 50% glycerol.


Upozornení: Reagent for in vitro laboratory use only.

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR