Tisk…

Storage plates, 96-well

Dodavatel: THERMO MATRIX TECHNOLOGIES
AB-0796
732-4885EA 5030 CZK
732-4885
Storage plates, 96-well
Mikrodestičky Destičky s hlubokými jamkami
PP. Ideal for compound storage, screening and culturing applications.

  • 96 round wells - maximum well volume 370 µl
  • U-shaped well bottom
  • Alphanumeric grid-referencing and industry standard footprint
  • Supporting ribs to aid stacking and improve centrifugation performance

Colour: Natural

Colour: Natural
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR