Tisk…
OmniTrays, Nunclon™Δ
Dodavatel: Thermo Fisher Scientific
165218
732-2602EA 11320 CZK
732-2602
OmniTrays, Nunclon™Δ
Mikrodestičky Destičky pro bunečné kultury
PS, sterile, single well OmniTrays with lid are ideal for a variety of uses.
  • Standard microplate format with notched corners make them suitable for robotic handling
  • Certified surface treatment for optimal cell growth and attachment
Culture area: 84 cm²
External dimensions: 128×86 mm