Tisk…
PCR microtubes and strips
Dodavatel: BIOTIX
PCT-0002-8C
732-2463EA 0 CZK
732-2463
PCR microtubes and strips
Zkumavky Zkumavky pro PCR
Medical grade virgin polypropylene.
  • Excellent chemical and temperature resistance
  • Biologically inert
  • BioReady™ certified free from DNase, RNase and endotoxin
Superior clarity for sample visualisation.