Tisk…
Single- and multi-well reservoirs, Axygen®
Dodavatel: Corning
RES-MW12-HP RES-MW12HPSI RES-MW12-LP RES-MW8-HP RES-MW8HPSI RES-MW8-LP RES-SW12-HP RES-SW12HPSI RES-SW1-LP RES-SW1LPSI RESSW384HPSI RES-SW384-LP RESSW384LPSI RES-SW8-HP RES-SW8-HPSI RES-SW96-LP RES-SW96LPSI RES-SW384-FX RES-SW384-FXSI RES-MW4-HP RES-MW8-LP-SI RES-MW12-LP-SI RES-SW8-HP-S RES-SW96-HPSIM
732-1390EA 10150 CZK
732-1390 613-2304 732-1391 732-1392 613-2306 732-1393 732-1394 AXYGRES-SW12HPSI 732-1395 AXYGRES-SW1LPSI AXYGRESSW384HPSI 732-1398 AXYGRESSW384LPSI 732-1399 AXYGRES-SW8-HPSI 732-1401 AXYGRES-SW96LPSI 732-1396 AXYGRES-SW384-FXSI AXYGRES-MW4-HP 613-2308 613-2307 AXYGRES-SW8-HP-S AXYGRES-SW96-HPSIM
Single- and multi-well reservoirs, Axygen®
Zásobníky na reagencie
These polypropylene reagent vessels are appropriate for automated and manual applications
  • Reservoirs are made of clear virgin polypropylene for excellent chemical resistance
  • All reservoirs feature an SBS footprint for compatibility with most robotic systems
  • Flat-bottom reservoirs are single-well format and are guaranteed flat
Trough reservoirs incorporate flow troughs that hold the sample across the length or breadth of the reservoir in either 8- or 12-trough configurations. Diamond-bottom reservoirs feature 96- and 384-well profiles that are idealMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Certifikát ochrany životního prostředí I-RECs, ISO 14001 - Reynosa I-RECs, ISO 14001 - Reynosa I-RECs, ISO 14001 - Reynosa I-RECs, ISO 14001 - Reynosa