Tisk…
Filter tips, ART® reload system
Dodavatel: Thermo Fisher Scientific
2065E-HR 2065E-RI 2065-HR 2065-RI 2069-RI 2139-RI 2140-RI 2149E-RI 2149P-RI 2179-RI 2065E-05-HR 2065E-05-RI 2069-05-RI 2139-05-RI 2140-05-RI 2149E-05-RI 2149P-05-RI 2179-05-RI 2931-RI
732-1144EA 0 CZK
732-1144 732-1152 732-1143 732-1151 732-1153 732-1147 732-1148 732-1149 732-1150 732-1154 732-1168 732-1167 732-1165 732-1180 732-1178 732-1176 732-1174 732-1172 732-1502
Filter tips, ART® reload system
Špičky pipetovací
This sterile barrier tip reload system combines individually packed blister tubs made of recycled plastic and new hinged racks. The push-down blister tub easily inserts the ART® barrier tips into the hinged racks providing stackable space saving racks that offer easy, one-handed use. Empty hinged racks allow for easy conversion to the ART® Reload system.
  • Environmentally friendly packaging
  • ART® self-sealing barrier prevent cross-contamination
  • Tips are certified DNase-, RNase-, ATP- and pyrogen-free