Tisk…
Tip refill systems, Axygen®
Dodavatel: Corning
RFL-222-C RFL-300-C RFL-1200-C
732-0956EA 1450 CZK
732-0956 732-0957 732-1402
Tip refill systems, Axygen®
Špičky pipetovací
Transparent PP, autoclavable.
  • Easy to load refill racks
  • Certified RNase-, DNase- and endotoxin-free
  • Universal fit tips have a bevelled reference and graduated marks