Tisk…
PCR plates, 384-well, Thermo-Fast® 384
Dodavatel: Thermo Fisher Scientific
AB-1384/P AB-1384/Y AB-1384/R AB-1384/G AB-1384/B AB-1384 AB-1384/W BC-1384 BC-1384/W
ADVAAB-1384/PEA 9730 CZK
ADVAAB-1384/P 731-0294 731-0293 731-0292 ADVAAB-1384/B 732-0897 732-0899 ADVABC-1384 ADVABC-1384/W
PCR plates, 384-well, Thermo-Fast® 384
Mikrodestičky Destičky pro PCR
PP. Designed for high throughput PCR, this 384-well plate is compatible with the majority of 384 block PCR and sequencing instruments. The full skirt and rigid plate deck minimise distortion during PCR making it suitable for use with robotic systems.
  • Cut-away corners
  • Fully skirted for compatibility with automated systems
  • Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
40 µl maximum well volume, 25 µl working volume