Tisk…
Mini tube systems (MTS) caps, Axygen®
Dodavatel: Corning
MTS-12CP-C MTS-12CP-C-S MTS-8CP-C MTS-8CP-C-S
731-0221EA 3080 CZK
731-0221 731-0222 731-0223 731-0224
Mini tube systems (MTS) caps, Axygen®
Uzávěry Víčka na zkumavky
Axygen® mini tube systems are available in 8-cap or 12-cap strip format.
  • Molded-in condensation rings on the lid minimise cross contamination