Tisk…
Vertical electrophoresis systems, Owl™ P81 single-sided
Dodavatel: THERMO OWL SCIENTIFIC
P81
730-0311EA 35340 CZK
730-0311
Vertical electrophoresis systems, Owl™ P81 single-sided
Systémy elektroforetické Systémy pro vertikální gelovou elektroforézu
The Owl™ P81 is a compact, single sided vertical electrophoresis system designed for economical, quick protein runs.
  • Simple to use, fast, efficient and provides excellent results
  • Upper buffer chamber extends the length of the gel to distribute heat evenly across the whole gel surface
  • Side clamps press the glass plates against a silicone gasket to provide uniform pressure and form a leak proof seal, resulting in flat, even banding patterns
  • For cooler, faster runs the system includes a notched alumina ceramic plate, which may be used in place of the notched
More Product Information