Tisk…
Blood collection tubes with gel and clot activator, vacuum, Vacutest®
Dodavatel: KIMA SAS DI CHIARIN RENZO
10174 10020 10118 10176 10214 10060 10138 10178
720-2726EA 1320 CZK
720-2726 720-2722 720-2724 720-2727 720-2729 720-2723 720-2725 720-2728
Blood collection tubes with gel and clot activator, vacuum, Vacutest®
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use, size 40×20 mm, printed in three colours.
  • Available in 3 sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
  • 18 months
More Product Information