Tisk…
IV catheter, Introcan®
Dodavatel: B. Braun
4253302 4254074B 4252098B 4254090B 4254112B 4254139B 4254171B 4254210B 4254325B 4252071B 4252110B 4252160B 4252322B
720-2506EA 3290 CZK
720-2506 720-2507 720-2505 720-2508 720-2509 720-2510 720-2511 720-2512 720-2545 BRAU4252071B BRAU4252110B BRAU4252160B BRAU4252322B
IV catheter, Introcan®
Katetry
Conventional IV catheter designed for single-handed insertion. Push-off plate on catheter hub facilitates single-handed puncture procedures. Sharp backcut bevel for minimal puncture trauma.
  • Four fully embedded x-ray contrast lines
  • Hydrophobic blood stoppers and lock fitting
  • Colour-coded sizes
  • Latex free and PVC free, catheter made of FEP
Meets ISO standard 10555-1/5 — same product properties as the Introcan Safety® (with PUR catheter) excluding the integrated, self-activating safety device for preventing needlestick injuries and the associated risk ofMore Product Information