Tisk…
Biopsy cassettes, ActivFlo™ Routine I
Dodavatel: LEICA HISTOLOGY
39LC-500-1-L 39LC-500-1 39LC-500-9-L 39LC-500-4 39LC-500-4-L 39LC-500-5 39LC-500-5-L 39LC-500-6 39LC-500-6-L 39LC-500-7 39LC-500-7-L 39LC-500-8 39LC-500-3 39LC-500-9 39LC-500-2-L 39LC-500-8-L 39LC-500-3-L 39LC-500-10 39LC-500-10-L 39LC-500-11 39LC-500-11-L 39LC-500-2
720-2278EA 4080 CZK
720-2278 720-2277 720-2470 720-2459 720-2460 720-2461 720-2462 720-2463 720-2464 720-2465 720-2466 720-2467 720-2457 720-2469 720-2456 720-2468 720-2458 720-2451 720-2452 720-2453 720-2454 720-2455
Biopsy cassettes, ActivFlo™ Routine I
Kazety pro zalévání vzorků Kazety bioptické
ActivFlo Routine I cassettes are processing cassettes featuring a patented lateral vent design for maximum fluid exchange. The increase in fluid exchange results in less incidence of unfixed or under-processed tissues and faster fixation/processing of tissue specimens. These one piece cassettes have ‘frangible’ lids for easy removal. The cassettes are designed for optimal workflow in the Leica IP C Cassette Printer. They offer <1% printing error rate.
  • With 45° writing surface
  • Available in taped packaging for easy printer loading
  • Eleven colours in