Tisk…
Cytology funnels, CytoSep™ for StatSpin Cytofuge® 2 cytocentrifuge
Dodavatel: Simport Scientific
M968-1
720-1972EA 2340 CZK
720-1972
Cytology funnels, CytoSep™ for StatSpin Cytofuge® 2 cytocentrifuge
Nálevky Nálevky cytologické
CytoSep™ cytology funnels for the StatSpin Cytofuge® 2 cytocentrifuge allow cells to gently concentrate in a 7 mm diameter area on the slide, while supernatant is simultaneously absorbed by a filter card. Will accept 50 to 400 μl of sample. Produces cell mono layer presentations of excellent quality.