Tisk…
Tissue cassettes in QuickLoad™ sleeves, Slimsette™, M509SL
Dodavatel: Simport Scientific
M509-10SL M509-11SL M509-12SL M509-2SL M509-3SL M509-4SL M509-5SL M509-6SL M509-7SL M509-8SL M509-9SL
720-1421EA 8100 CZK
720-1421 720-1423 720-1425 720-1430 720-1432 720-1434 720-1436 720-1438 720-1440 720-1442 720-1444
Tissue cassettes in QuickLoad™ sleeves, Slimsette™, M509SL
Kazety pro zalévání vzorků Kazety tkáňové
Acetal polymer. This model is similar to the M507 cassette but is intended for tissue processing/embedding procedures.
  • Large labelling areas are located on three sides of the cassette
  • Anterior writing area at a 45° angle
  • Available in several non cytotoxic, non metallic colours
The transparent sleeve allows viewing of cassettes in order to confirm there is no jam in the sleeve during the printing process. The efficient flow-through slots maximise fluid exchange and ensure proper drainage. Lids can be opened and closed as often as necessary and theyMore Product Information