Tisk…
Biopsy cassettes in QuickLoad™ taped stacks, Unisette™, M506T
Dodavatel: Simport Scientific
M506-10T M506-11T M506-12T M506-2T M506-3T M506-4T M506-5T M506-6T M506-7T M506-8T M506-9T
720-1400EA 6870 CZK
720-1400 720-1401 720-1402 720-1403 720-1404 720-1405 720-1406 720-1407 720-1408 720-1409 720-1410
Biopsy cassettes in QuickLoad™ taped stacks, Unisette™, M506T
Kazety pro zalévání vzorků Kazety bioptické
Moulded from a special high density acetal polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.
  • The snap-latch and hinge-lock design prevents early separation of base and lid and allows for one handed operation
  • Large labelling areas are located on two sides of the cassettes
  • Available in several colours
The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. LidsMore Product Information