Tisk…
Staining racks, with dishes, WHEATON®
Dodavatel: DWK Life Sciences
900234 900254 900301 900302 900303 900304
720-1030EA 6900 CZK
720-1030 720-0702 720-0703 720-0704 720-0705 720-0706
Staining racks, with dishes, WHEATON®
Stojany Stojany na barviva
Soda-lime glass and stainless steel.
  • Rack for 16 standard slides (32 back to back)
  • Rack for 40 standard slides (76×26 mm) or for use with 76×38 mm slides or 76×51 mm slides
Dishes with lids and rack available in different sizes.
W×D×H: 70×120×90 mm