Tisk…
E-Z Load™ Histosette® II M482 & M483, E-Z Load™ QuickLoad™ M485SL & M486SL
Dodavatel: Simport Scientific
M486-6SL M486-5SL M485-10SL M485-11SL M485-12SL M485-2SL M485-3SL M485-4SL M485-6SL M486-10SL M486-11SL M486-12SL M486-2SL M486-3SL M486-4SL M485-5SL M483-11BA M483-6LI M483-11LI M483-12BA M483-12LI M483-2BA M483-2LI M483-3BA M483-3LI M483-4BA M483-4LI M483-5BA M483-6BA M483-7BA M483-7LI M483-8BA M483-8LI M483-9BA M483-9LI M483-5LI M482-6LI M482-11LI M482-12BA M482-12LI M482-2BA M482-2LI M482-3BA M482-3LI M482-4BA M482-4LI M482-5BA M482-6BA M482-10BA M482-7BA M482-7LI M482-8BA M482-8LI M482-9BA M482-9LI M483-10BA M483-10LI M482-5LI M482-11BA M482-10LI
720-1222EA 6190 CZK
720-1222 720-1221 720-1207 720-1208 720-1209 720-1210 720-1211 720-1212 720-1214 720-1215 720-1216 720-1217 720-1218 720-1219 720-1220 720-1213 720-0864 720-0877 720-0865 720-0866 720-0867 720-0868 720-0869 720-0870 720-0871 720-0872 720-0873 720-0874 720-0876 720-0878 720-0879 720-0880 720-0881 720-0882 720-0883 720-0875 720-0855 720-0843 720-0844 720-0845 720-0846 720-0847 720-0848 720-0849 720-0850 720-0851 720-0852 720-0854 720-0840 720-0856 720-0857 720-0858 720-0859 720-0860 720-0861 720-0862 720-0863 720-0853 720-0842 720-0841
E-Z Load™ Histosette® II M482 & M483, E-Z Load™ QuickLoad™ M485SL & M486SL
Kazety pro zalévání vzorků Kazety do štítkovacích automatů
Acetal polymer. E-Z Load™ Histosette® II cassettes are designed for loading Micro Writer® and Shur/Mark® cassette labelling instruments in one simple operation. Supplied in stacks of 50 cassettes, no manual insertion, one cassette at a time, is required. Just load the stack in the magazine, cut and remove the holding tie and place in the labelling instrument ready for printing. Moulded from a special high density polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slotsMore Product Information