Tisk…
Embedding cassettes, M480
Dodavatel: Simport Scientific
M480-2 M480-9 M480-11 M480-12 M480-3 M480-4 M480-5 M480-6 M480-7 M480-8 M480-1 M480-10
720-0832EA 5390 CZK
720-0832 720-0839 720-0830 720-0831 720-0833 720-0834 720-0835 720-0836 720-0837 720-0838 720-0828 720-0829
Embedding cassettes, M480
Kazety pro zalévání vzorků Kazety tkáňové
Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures.
  • Resistant to all commonly used histological solvents
  • Designed to accept standard metal lids (M481)
  • Slanted writing surface (45° angle) accepts markings easily
Different colours available.