Tisk…

Silikagel, Supelco®

Dodavatel: Merck

717185-1KG 717185-25KG 717185-5KG
717185-1KGEA 4010 CZK
717185-1KG 717185-25KG 717185-5KG
Silikagel, Supelco®
Silikagel
Vzorec: SiO₂
Bod varu: 2230 °C (1013 hPa)
Bod tání: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS číslo: 63231-67-4
REACH: 01-2119379499-16
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=991ad213ed142a5909207736ee16a0ae85014942&vid=4599672e7742c89c4f618eaf4db6311e437a5fb8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a2419c5763bb40cef2478bcba336dd641cabf7f&vid=e73855d1ffa4caa2b074b1d7daeefea6254aa146&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dbdd5a5146a5a24998f98bc7da8ff8ea425e462&vid=022b518cfd591a8c543ede89f2baefa0ad11d9bf&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR