Tisk…

Dibutyltin dilaurate ≥95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Di-n-butyltin dilaurate

71130.22 71130.36 71130.14
71130.22EA 1980 CZK
71130.22 71130.36 71130.14
Dibutyltin dilaurate ≥95%
Dibutyltin dilaurate

Purity: 95% CAS No.: 77-58-7 Molecular Formula: C32H64O4Sn Formula Weight: 631.55 Flash Point: 235°(455°F) Physical Form: Liquid Density: 1.066 Refractive Index: 1.4730 Appearance: Pale yellow Melting Point: 10° Boiling Point: 204°/12mm Solubility: Soluble in benzene and ether. Insoluble in water and methanol Merck Reference: 14,3038 UN No.: 2788 MDL No.: MFCD00008963

Vzorec: C₃₂H₆₄O₄Sn
MW: 631.57
Bod varu: >204 °C (12 mmHg)
Hustota: 1.066
Bod vzplanutí: 235 °C (455 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008963
CAS číslo: 77-58-7
EINECS: 201-039-8
UN: 2788
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR