Tisk…
Secchi disc
Dodavatel: Hach
2177200
707-0005EA 4490 CZK
707-0005
Secchi disc
Zákalometry
The disc is designed to measure water transparency in situ via the semi-quantitative method.
  • Disc slowly lowered in the water until the pattern is no longer visible
  • Depth of immersion is then evaluated
  • Measure related to the transparency
For use in oceans and lakes.
Ø = 200 mm
Certifikace: ISO 7027