Tisk…

Multiparameter Handheld photometers, Lovibond® MD 100

Dodavatel: Lovibond

278060 278070 278080 278090 278000 278010 278020 278025 278030
705-0677EA 20540 CZK
705-0677 705-0678 705-0679 705-0674 705-0675 705-0676 LOVI278020 LOVI278025 LOVI278030
Multiparameter Handheld photometers, Lovibond® MD 100
Kity pro environmentální testy

The MD 100 range of photometers are portable handheld instruments with a simple button layout, robust waterproof design and large backlit display that are ideal for testing in the field or laboratory conditions.


  • Accurate (3% FS), reproducible results using high quality interference filters with long-life LEDs
  • Scroll memory - remembers the last test method before switching off
  • Real time clock and date
  • OTZ (One Time Zero setting) on versions except those denoted*
  • Optional infrared interface for transfer of test data to PC or printer


Informace o dodávce: Instrument supplied with carry case, 4× AAA batteries, 3× round glass vials with lids, relevant reagents, and instruction manual.


Upozornení: * OTZ not applicable

Objednat

Produktová specifikace

Model MD 100 MD 100 MD 100 MD 100
Environmentally Preferable
Přesnost 3% FS (20...25 °C) 3% FS (20...25 °C) 3% FS (20...25 °C) 3% FS (20...25 °C)
Vlnové délky 430, 530, 560, 580, 610, 660 nm - selection depending on the version 430, 530, 560, 580, 610, 660 nm - selection depending on the version 430, 530, 560, 580, 610, 660 nm - selection depending on the version 430, 530, 560, 580, 610, 660 nm - selection depending on the version
Přesnost vlnové délky ±1 nm ±1 nm ±1 nm ±1 nm
Životnost baterie 17 h or 5000 tests 17 h or 5000 tests 17 h or 5000 tests 17 h or 5000 tests
Displej LCD (backlit on keypress) LCD (backlit on keypress) LCD (backlit on keypress) LCD (backlit on keypress)
Rozhraní Infrared interface for data transfer Infrared interface for data transfer Infrared interface for test data transfer Infrared interface for test data transfer
Numerická klávesnice 4 button membrane keypad 4 button membrane keypad 4 button membrane keypad 4 button membrane keypad
Zdroj světla LED LED LED LED
Provozní vlhkost 30 - 90% RH 30 - 90% RH 30 - 90% RH 30 - 90% RH
Provozní teplota 5...40 °C 5...40 °C 5...40 °C 5...40 °C
Optika LED, interference filters (IF) and photo sensor in transparent sample chamber LED, interference filters (IF) and photo sensor in transparent sample chamber LED, interference filters (IF) and photo sensor in transparent sample chamber LED, interference filters (IF) and photo sensor in transparent sample chamber
Napájení 4× AAA batteries 4× AAA batteries 4× AAA batteries 4× AAA batteries
Úložná kapacita Internal ring memory for 16 data sets Internal ring memory for 16 data sets Internal ring memory for 16 data sets Internal ring memory for 16 data sets
Hmotnost ~0,26 kg ~0,26 kg ~0,26 kg ~0,26 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR