Tisk…
Kjeldahl tablets, Kjeltabs
Dodavatel: THOMPSON & CAPPER
AA00 AA08 AA09 AA10 AA11 AA13 AA15 AA16 AA17 AA18 AA20 AA21 AA22 AA25 AA41 AA44 AA47 AA48 AA52
705-0543EA 12880 CZK
705-0543 705-0510 705-0511 705-0512 705-0545 705-0513 705-0514 705-0515 705-0516 705-0517 705-0518 705-0519 705-0546 705-0520 705-0547 705-0548 705-0549 THOMAA48 705-0521
Kjeldahl tablets, Kjeltabs
Systémy rozkladové Kjeldahl - tablety a činidla
Kjeldahl catalyst reagents in a convenient tablet form with a range of tablets to suit general and specific applications.
Note: Most of these materials are hazardous. Please read the material safety data sheets prior to use.