Tisk…
SpecCheck secondary gel standards sets
Dodavatel: Hach
2635300 2712500 2708000 2893300 2507500
705-0167EA 5240 CZK
705-0167 HACH2712500 HACH2708000 HACH2893300 HACH2507500
SpecCheck secondary gel standards sets
Standardy Standardy pro environmentální analýzu
  • Convenient - check instrument response without sample or reagent preparation
  • Coloured gels are stable - assuring consistent performance