Tisk…
Scrubbers, K-415 series
Dodavatel: BUCHI
114152331 114152320 114152330 114152341
701-0238EA 232070 CZK
701-0238 701-0236 701-0237 701-0239
Scrubbers, K-415 series
Destilační systémy Mycí systémy
A modular, upgradeable, system for scrubbing of harmful fumes from distillation or digestion processes. The system can be obtained with up to four process stages depending on user requirements.
DuoScrub: Basic, two stage model with a neutralisation vessel and an absorption vessel linked to a powerful suction pump (140 W, up to 32 L/min.).
TripleScrub and TripleScrubECO: Three stage model with pre-condensation step as stage 1 to collect entrained liquids and avoid dilution of the neutralisation stage 2. The TripleScrubECO model is equipped with automatic cooling water control andMore Product Information