Tisk…
Test strips, drug testing for urine
Dodavatel: SURESCREEN
AMS BZS CAS MTDS OPS2000 TRAMS
700-9544EA 2350 CZK
700-9544 720-0657 700-9564 720-0737 700-9634 SURETRAMS
Test strips, drug testing for urine
Testy na drogy
Single test strips are an efficient method for testing urine for specific drugs in urine. Simply dip the test strip into a urine sample and read at 3 minutes. Each individual strip is provided in a foil pouch, to ensure the high quality of the product is retained throughout the shelf-life.
  • More than 20 different drug tests available
  • Easy to perform test routine
  • Clear results in 3 minutes
  • Greater than 99% accuracy
  • For detecting recent drug use approx. 1 to 8 days
Recommended markets and use:
On-site screening
Safety critical situations
Random screening
More Product Information