Tisk…

Dimethyl selenide

Dodavatel: Thermo Fisher Scientific
Nebezpečí

68116.06 68116.14 68116.03
68116.06EA 7210 CZK
68116.06 68116.14 68116.03
Dimethyl selenide
Dimethyl selenide

CAS No.: 593-79-3 Molecular Formula: C2H6Se Formula Weight: 109.03 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.41 Appearance: Colourless Odour: Unpleasant Boiling Point: 56-58° Beilstein Reference: 1696848 UN No.: 3282 MDL No.: MFCD00014848

Vzorec: C₂H₆Se
Bod varu: 56…58 °C
Hustota: 1,41
Storage Temperature: Refrigerator
MDL Number: MFCD00014848
CAS číslo: 593-79-3
EINECS: 209-807-4
UN: 3282
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR