Tisk…

Scandium(III) nitrate hydate ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trinitric acid scandium salt hydrate, Scandium(3+) trinitrate hydrate, Scandium trinitrate hydrate

67104.03 67104.06
67104.03EA 4330 CZK
67104.03 67104.06
Scandium(III) nitrate hydate ≥99.9% (REO, rare earth oxide basis), REacton®
Scandium(III) nitrate hydrate

Purity: 99.9% CAS No.: 13465-60-6 Molecular Formula: Sc(NO3)3.xH2O Formula Weight: 230.97(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Solubility: Freely soluble in water and alcohol Merck Reference: 14,8392 UN No.: 1477 MDL No.: MFCD00011220

Vzorec: Sc(NO₃)₃·xH₂O
MW: 230.97 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011220
CAS číslo: 107552-14-7
EINECS: 236-701-5
UN: 1477
ADR: 5.1 III
Merck Index: 14,8392

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR