Tisk…
pH combination electrodes, IntelliCAL™, High Performance, Red Rod
Dodavatel: Hach
PHC73501 PHC74501 PHC70501 PHC705A01 PHC72501 PHC72901 PHC80501
665-0388EA 21420 CZK
665-0388 665-0389 665-0384 665-0385 665-0386 665-0387 665-0390
pH combination electrodes, IntelliCAL™, High Performance, Red Rod
Elektrody Elektrody pH
Digital high-performance pH electrodes with Red Rod reference system and built-in temperature sensor.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Red Rod system creates a trouble-free liquid junction
  • Encapsulation of reference elements: no silver ions and therefore less risk of clogging when measuring in challenging sample matrices
  • Electrodes store time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • Electrodes can easily be moved between different HQD meters
  • Ideal for a