Tisk…
Sondy pro rozpuštěný kyslík, IDS, FDO®
Dodavatel: WTW
201301 201300 201306
664-0051EA 34010 CZK
664-0051 664-0050 665-0303
Sondy pro rozpuštěný kyslík, IDS, FDO®
Elektrody Sondy pro stanovení kyslíku
Optical dissolved oxygen probes for field and laboratory applications. For use with IDS meters.
  • Extremely fast and precise
  • Factory calibrated cap with built-in data chip
  • Cables are reinforced with Kevlar® which ensures safe measurements in deep lakes or torrential rivers
  • Slender design (suitable for Winkler and Karlsruher bottles)
  • Bevelled membrane repels interference that can be caused by air bubbles and allows the sensors to be used in containers with low sample volumes
Ideal for wastewater treatment, BSB measurements in Karlsruhe bottles and duringMore Product Information