Tisk…
Multiparameter photometer, Water-ID® The PrimeLab 2.0
Dodavatel: WATER ID PRIMELAB
PL02BALL PL02BALL-WO90 PL02BALL-COD ELECHRGLATCKIT ELECHRPCATCKIT ELECLRGL-KIT ELECLRPCATCKIT ELORPPCATC-KIT ELPHGLATC-KIT ELPHGLREFATCKT ELPHPCATC-KIT ELECHRGLATC ELECHRPCATC PL2SP-ELECLRGL ELECLRPCATC ELORPPCATC ELPHGLATC ELPHGLREFATC -ELPHPCATC
665-0562EA 0 CZK
665-0562 665-0563 664-0241 663-0376 663-0378 663-0380 663-0382 664-0240 662-2428 662-2430 662-2432 663-0375 663-0377 663-0379 663-0381 664-0239 662-2427 662-2429 662-2431
Multiparameter photometer, Water-ID® The PrimeLab 2.0
Kolorimetry Kolorimetry víceparametrové
The next generation of multitest Photometer, with pH - EC - TDS - ORP - Temperature measurement functionality.
  • More than 140 parameters
  • 18 wavelengths (410 to 940 nm)
  • 5.5" colour, HD touch display
  • In-build barcode/QR-code scanner
  • USB Type–C (for charging, data transfer, probe connection and 4G modem)
  • Modern connection - WiFi, USB, 4G, Bluetooth®
  • Probe connector (pH, EC, TDS, Temp and ORP)
  • Auto-calibration function with certificate (software)
High-precision readings due to 18 parallel wavelengths, WiFi–USB–Bluetooth®–GSM connection, powerful softwareMore Product Information