Tisk…
Conductivity/°C meters, bench, inoLab® Cond 7110 / 7310
Dodavatel: WTW
1CA100 1CA101 1CA300P 1CA301 1CA300
663-0167EA 33060 CZK
663-0167 663-0168 663-0171 663-0172 663-0170
Conductivity/°C meters, bench, inoLab® Cond 7110 / 7310
Meřiče elektrochemické Konduktometry
High quality instruments with advanced measuring technology, making measurements more convenient and reliable than ever.
inoLab® Cond 7110: Accurate measurements
The inoLab® 7110 series is perfectly suited for routine measurements in general laboratory applications. Whether measuring conductivity in environmental, chemical, pharmaceutical, medical or food and beverage industries, this series offers advanced features in a simple to use package.
- AutoRead function ensures repeatable results
- Easy calibration including calibration timer
- Intuitive user-interface, large display and keypadMore Product Information