Tisk…
pH electrodes, InLab® Power
Dodavatel: Mettler - Toledo
51343110
662-2845EA 15170 CZK
662-2845
pH electrodes, InLab® Power
Elektrody Elektrody pH
Combination pH electrode with SteadyForce™ and S7 screw head for general laboratory applications.
  • Highly reproducible
  • Maintenance-free
  • Autoclavable
Informace o dodávce: Supplied with quality certificate.